Learn More
Meso Scale Discovery R-PLEX Human MIC-A ASY
SDP
Catalog No. 5000079994
Encompass
The R-PLEX Human MIC A Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that allows simultaneous quantitative measurement of IL-1 IL-1 IL-1ra IL-18 IL-33 and IL-37 in serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The assay requires 50 L of sample and can be completed in under 5 hours With minimal hands-on time this multiplex panel enables efficient analysis of multiple related human inflammatory biomarkers
Catalog No. 50-000-79994 Supplier Meso Scale Discovery Supplier No. K151L5R2
The R-PLEX Human MIC A Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that allows simultaneous quantitative measurement of IL-1 IL-1 IL-1ra IL-18 IL-33 and IL-37 in serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The assay requires 50 L of sample and can be completed in under 5 hours With minimal hands-on time this multiplex panel enables efficient analysis of multiple related human inflammatory biomarkers