Learn More
Meso Scale Discovery R-PLEX Human MIC-A ASYQP
SDP
Catalog No. 5000079995
Encompass
The R-PLEX Human MIC A Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that allows simultaneous quantitative measurement of IL-1 IL-1 IL-1ra IL-18 IL-33 and IL-37 in serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and assessing assay performance This assay allows efficient profiling of multiple inflammatory cytokines involved in innate immunity and inflammation
Catalog No. 50-000-79995 Supplier Meso Scale Discovery Supplier No. K151L5R22
Add to Cart
Add to Cart
The R-PLEX Human MIC A Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that allows simultaneous quantitative measurement of IL-1 IL-1 IL-1ra IL-18 IL-33 and IL-37 in serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and assessing assay performance This assay allows efficient profiling of multiple inflammatory cytokines involved in innate immunity and inflammation