Learn More
Meso Scale Discovery R-PLEX Human MIC-B ASY
SDP
Catalog No. 5000081978
Encompass
The R-PLEX Human MIC-B Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring biomarkers of infection and inflammation It quantifies human CRP SAA PCT LBP sCD14 and sCD163 in serum plasma and other samples Using electrochemiluminescent detection it offers high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies read buffer wash buffer calibrator and controls aiding in diagnosing bacterial and viral infections
Catalog No. 50-000-81978 Supplier Meso Scale Discovery Supplier No. K151AMDR2
The R-PLEX Human MIC-B Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring biomarkers of infection and inflammation It quantifies human CRP SAA PCT LBP sCD14 and sCD163 in serum plasma and other samples Using electrochemiluminescent detection it offers high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies read buffer wash buffer calibrator and controls aiding in diagnosing bacterial and viral infections