Learn More
Meso Scale Discovery R-PLEX Human Midkine ASY
SDP
Catalog No. 5000081931
Encompass
The R-PLEX Human Midkine Assay is a multiplex immunoassay kit for measuring Midkine in human serum plasma urine cell culture supernatants and other samples It uses Meso Scale Discovery s electrochemiluminescent detection technology offering high sensitivity broad dynamic range and precision with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls for efficient Midkine quantification With a range of 1 53 - 10 000 pg/mL it enables accurate analysis of Midkine levels for research
Catalog No. 50-000-81931 Supplier Meso Scale Discovery Supplier No. K151ALDR2
The R-PLEX Human Midkine Assay is a multiplex immunoassay kit for measuring Midkine in human serum plasma urine cell culture supernatants and other samples It uses Meso Scale Discovery s electrochemiluminescent detection technology offering high sensitivity broad dynamic range and precision with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls for efficient Midkine quantification With a range of 1 53 - 10 000 pg/mL it enables accurate analysis of Midkine levels for research