Learn More
Meso Scale Discovery R-PLEX Human Midkine ASYQP
SDP
Catalog No. 5000079565
Encompass
The R-PLEX Human Midkine Assay is a multiplex immunoassay kit by Meso Scale Diagnostics that allows for the quantitative measurement of human Midkine in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Diagnostics QuickPlex SQ 120 instrument The kit includes a plate coated with capture antibodies detection antibodies calibrators and diluent Results are obtained in as little as 3 5 hours
Catalog No. 50-000-79565 Supplier Meso Scale Discovery Supplier No. K151ALDR22
The R-PLEX Human Midkine Assay is a multiplex immunoassay kit by Meso Scale Diagnostics that allows for the quantitative measurement of human Midkine in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Diagnostics QuickPlex SQ 120 instrument The kit includes a plate coated with capture antibodies detection antibodies calibrators and diluent Results are obtained in as little as 3 5 hours