Learn More
Meso Scale Discovery R-PLEX Human MIP-4 ASYQP
SDP
Catalog No. 5000080555
Encompass
The R-PLEX Human MIP-4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that enables simultaneous quantification of the chemokine MIP-4 in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure sample concentrations The assay has a typical lower limit of detection of 0 04 pg/mL and an upper limit of quantitation of 10 000 pg/mL for MIP-4
Catalog No. 50-000-80555 Supplier Meso Scale Discovery Supplier No. K1512HR22
The R-PLEX Human MIP-4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that enables simultaneous quantification of the chemokine MIP-4 in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure sample concentrations The assay has a typical lower limit of detection of 0 04 pg/mL and an upper limit of quantitation of 10 000 pg/mL for MIP-4