Learn More
Meso Scale Discovery R-PLEX Human MMP-13 ASY
SDP
Catalog No. 5000081619
Encompass
The R-Plex Human MMP-13 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of human Matrix Metalloproteinase 13 (MMP-13) in serum plasma cell culture supernatants cell lysates tissue homogenates and other biological fluids It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be used to study MMP-13 levels in research areas like arthritis cancer cardiovascular disease and tissue remodeling With minimal hands-on time this kit allows for rapid profiling of MMP-13 across multiple samples
Catalog No. 50-000-81619 Supplier Meso Scale Discovery Supplier No. K151AJSR2
Add to Cart
Add to Cart
The R-Plex Human MMP-13 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of human Matrix Metalloproteinase 13 (MMP-13) in serum plasma cell culture supernatants cell lysates tissue homogenates and other biological fluids It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be used to study MMP-13 levels in research areas like arthritis cancer cardiovascular disease and tissue remodeling With minimal hands-on time this kit allows for rapid profiling of MMP-13 across multiple samples