Learn More
Meso Scale Discovery R-PLEX Human MMP-3 ASY
SDP
Catalog No. 5000079665
Encompass
The R-PLEX Human MMP-3 Assay Kit from Meso Scale Discovery is used to quantitatively measure MMP-3 levels in human serum plasma saliva urine cell culture supernatant and other biological samples It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for active MMP-3 with no cross-reactivity to pro-MMP-3 The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrator It can detect MMP-3 levels between 0 03-1000 ng/mL
Catalog No. 50-000-79665 Supplier Meso Scale Discovery Supplier No. K151ZFR2
The R-PLEX Human MMP-3 Assay Kit from Meso Scale Discovery is used to quantitatively measure MMP-3 levels in human serum plasma saliva urine cell culture supernatant and other biological samples It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for active MMP-3 with no cross-reactivity to pro-MMP-3 The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrator It can detect MMP-3 levels between 0 03-1000 ng/mL