Learn More
Meso Scale Discovery R-PLEX Human MMP-8 ASY
SDP
Catalog No. 5000079459
Encompass
The R-PLEX Human MMP-8 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures MMP-8 protein levels in human serum plasma saliva and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 1500 ng/mL
Catalog No. 50-000-79459 Supplier Meso Scale Discovery Supplier No. K1519ER2
The R-PLEX Human MMP-8 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures MMP-8 protein levels in human serum plasma saliva and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 1500 ng/mL