Learn More
Meso Scale Discovery R-PLEX Human MMP-8 ASYQP
SDP
Catalog No. 5000079026
Encompass
The R-PLEX Human MMP-8 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures levels of matrix metalloproteinase 8 (MMP-8) in human serum plasma saliva and cell culture supernatant samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 ng/mL
Catalog No. 50-000-79026 Supplier Meso Scale Discovery Supplier No. K1519ER22
The R-PLEX Human MMP-8 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures levels of matrix metalloproteinase 8 (MMP-8) in human serum plasma saliva and cell culture supernatant samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 ng/mL