Learn More
Meso Scale Discovery R-PLEX Human MPIF-1/CCL23 ASY
SDP
Catalog No. 5000080169
Encompass
The R-PLEX Human MIF-1 (CCL23) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the chemokine Macrophage Migration Inhibitory Factor (MIF-1 or CCL23) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator and sample diluent The assay can detect MIF-1 concentrations as low as 3 5 pg/mL with an upper limit of 100 000 pg/mL
Catalog No. 50-000-80169 Supplier Meso Scale Discovery Supplier No. K151G1R2
The R-PLEX Human MIF-1 (CCL23) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the chemokine Macrophage Migration Inhibitory Factor (MIF-1 or CCL23) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator and sample diluent The assay can detect MIF-1 concentrations as low as 3 5 pg/mL with an upper limit of 100 000 pg/mL