Learn More
Meso Scale Discovery R-PLEX Human MPO ASY
SDP
Catalog No. 5000078825
Encompass
The R-PLEX Human MPO Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of myeloperoxidase (MPO) in human serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide high sensitivity wide dynamic range and low sample volume requirements The assay has a lower limit of detection of 0 04 ng/mL and an upper limit of 1000 ng/mL The total assay time is 3 5 hours This product can aid in the evaluation of inflammatory conditions where MPO is a biomarker
Catalog No. 50-000-78825 Supplier Meso Scale Discovery Supplier No. K1514ER2
The R-PLEX Human MPO Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of myeloperoxidase (MPO) in human serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide high sensitivity wide dynamic range and low sample volume requirements The assay has a lower limit of detection of 0 04 ng/mL and an upper limit of 1000 ng/mL The total assay time is 3 5 hours This product can aid in the evaluation of inflammatory conditions where MPO is a biomarker