Learn More
Meso Scale Discovery R-PLEX Human MPO ASYQP
SDP
Catalog No. 5000078826
Encompass
The R-PLEX Human MPO Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures myeloperoxidase (MPO) levels in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay utilizes MSD s proprietary SECTOR Imager and provides a sensitive and reproducible method for quantifying MPO with a detection range of 0 04-500 ng/mL and an LLOQ of 0 04 ng/mL The assay requires 50 L of sample and can be completed in under 5 hours This kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent It provides a high-throughput and cost-effective tool for investigating MPO levels in research studies
Catalog No. 50-000-78826 Supplier Meso Scale Discovery Supplier No. K1514ER22
The R-PLEX Human MPO Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures myeloperoxidase (MPO) levels in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay utilizes MSD s proprietary SECTOR Imager and provides a sensitive and reproducible method for quantifying MPO with a detection range of 0 04-500 ng/mL and an LLOQ of 0 04 ng/mL The assay requires 50 L of sample and can be completed in under 5 hours This kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent It provides a high-throughput and cost-effective tool for investigating MPO levels in research studies