Learn More
Meso Scale Discovery R-PLEX Human MSP ASY
SDP
Catalog No. 5000081979
Encompass
The R-PLEX Human MSP Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures Macrophage Stimulating Protein (MSP) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and is provided in a 96-well plate coated with capture antibodies The kit includes a calibrator and controls to generate a standard curve and validate assay performance This quantitative assay has high sensitivity requires a small sample volume and provides results in as little as 3 5 hours With minimal hands-on time required this kit allows for the efficient analysis of MSP across large sample sets
Catalog No. 50-000-81979 Supplier Meso Scale Discovery Supplier No. K151AMER2
The R-PLEX Human MSP Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures Macrophage Stimulating Protein (MSP) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and is provided in a 96-well plate coated with capture antibodies The kit includes a calibrator and controls to generate a standard curve and validate assay performance This quantitative assay has high sensitivity requires a small sample volume and provides results in as little as 3 5 hours With minimal hands-on time required this kit allows for the efficient analysis of MSP across large sample sets