Learn More
Meso Scale Discovery R-PLEX Human MSP ASYQP
SDP
Catalog No. 5000078331
Encompass
The R-PLEX Human MSP Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures Macrophage Stimulating Protein (MSP) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and is provided in a 96-well plate coated with capture antibodies The kit includes a calibrator and controls to generate a standard curve and validate assay performance MSP is measured with high sensitivity and the small sample volume required makes this assay ideal for precious clinical samples This quantitative immunoassay delivers rapid results using an automated plate reader
Catalog No. 50-000-78331 Supplier Meso Scale Discovery Supplier No. K151AMER22
The R-PLEX Human MSP Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures Macrophage Stimulating Protein (MSP) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescence detection technology and is provided in a 96-well plate coated with capture antibodies The kit includes a calibrator and controls to generate a standard curve and validate assay performance MSP is measured with high sensitivity and the small sample volume required makes this assay ideal for precious clinical samples This quantitative immunoassay delivers rapid results using an automated plate reader