Learn More
Meso Scale Discovery R-PLEX Human MyD88 ASY
SDP
Catalog No. 5000081296
Encompass
The R-PLEX Human MyD88 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures MyD88 protein levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator The assay has a typical sensitivity of 19 pg/mL and an upper limit of quantification of 50 000 pg/mL It can be used to monitor MyD88 expression and signaling in immunology and oncology research
Catalog No. 50-000-81296 Supplier Meso Scale Discovery Supplier No. K151S4R2
The R-PLEX Human MyD88 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures MyD88 protein levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator The assay has a typical sensitivity of 19 pg/mL and an upper limit of quantification of 50 000 pg/mL It can be used to monitor MyD88 expression and signaling in immunology and oncology research