Learn More
Meso Scale Discovery R-PLEX Human Myoglobin ASY
SDP
Catalog No. 5000079429
Encompass
The R-PLEX Human Myoglobin Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of myoglobin in human serum plasma and urine It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results This assay allows for the simultaneous measurement of myoglobin and other biomarkers from a single small sample volume It includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls The kit has a measuring range of 50-50 000 pg/mL and a lower limit of detection of 17 pg/mL for myoglobin It is intended for research use only
Catalog No. 50-000-79429 Supplier Meso Scale Discovery Supplier No. K151AAZR2
The R-PLEX Human Myoglobin Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of myoglobin in human serum plasma and urine It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results This assay allows for the simultaneous measurement of myoglobin and other biomarkers from a single small sample volume It includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls The kit has a measuring range of 50-50 000 pg/mL and a lower limit of detection of 17 pg/mL for myoglobin It is intended for research use only