Learn More
Meso Scale Discovery R-PLEX Human Myoglobin ASYQP
SDP
Catalog No. 5000079615
Encompass
The R-PLEX Human Myoglobin Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring myoglobin in human serum plasma and urine It uses electrochemiluminescent detection for high sensitivity and broad dynamic range with low sample volume The detection limit is 0 8 ng/mL and the quantification limit is 10 000 ng/mL The kit includes a 96-well plate labeled detection antibodies calibrators controls buffers and seals It is compatible with SECTOR Imager instruments for automated analysis
Catalog No. 50-000-79615 Supplier Meso Scale Discovery Supplier No. K151AAZR22
The R-PLEX Human Myoglobin Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring myoglobin in human serum plasma and urine It uses electrochemiluminescent detection for high sensitivity and broad dynamic range with low sample volume The detection limit is 0 8 ng/mL and the quantification limit is 10 000 ng/mL The kit includes a 96-well plate labeled detection antibodies calibrators controls buffers and seals It is compatible with SECTOR Imager instruments for automated analysis