Learn More
Meso Scale Discovery R-PLEX Human N-Cadherin ASY
SDP
Catalog No. 5000078902
Encompass
The R-Plex Human N-Cadherin Assay is a multiplex immunoassay kit for measuring N-Cadherin in human serum plasma and cell culture supernatants It uses a quantitative sandwich enzyme immunoassay and color-coded microspheres to measure multiple biomarkers in one well The kit includes microplates detection antibodies standards controls diluents wash solutions substrates and stop solutions for 96 tests With a range of 1 56 - 400 ng/mL and sensitivity of 0 64 ng/mL it ensures sensitive reproducible and accurate quantification of N-Cadherin levels
Catalog No. 50-000-78902 Supplier Meso Scale Discovery Supplier No. K151ABTR2
The R-Plex Human N-Cadherin Assay is a multiplex immunoassay kit for measuring N-Cadherin in human serum plasma and cell culture supernatants It uses a quantitative sandwich enzyme immunoassay and color-coded microspheres to measure multiple biomarkers in one well The kit includes microplates detection antibodies standards controls diluents wash solutions substrates and stop solutions for 96 tests With a range of 1 56 - 400 ng/mL and sensitivity of 0 64 ng/mL it ensures sensitive reproducible and accurate quantification of N-Cadherin levels