Learn More
Meso Scale Discovery R-PLEX Human N-Cadherin ASYQP
SDP
Catalog No. 5000079453
Encompass
The R-Plex Human N-Cadherin Assay is a multiplex immunoassay kit for measuring N-Cadherin in human serum plasma and cell culture supernatants It uses a quantitative sandwich enzyme immunoassay and color-coded microspheres to measure multiple biomarkers in one well The kit includes microplates detection antibodies standards controls diluents wash solutions substrates and stop solutions for 96 tests With a range of 1 56-400 ng/mL and minimal sample volume it allows rapid reproducible quantification of N-Cadherin across samples
Catalog No. 50-000-79453 Supplier Meso Scale Discovery Supplier No. K151ABTR22
The R-Plex Human N-Cadherin Assay is a multiplex immunoassay kit for measuring N-Cadherin in human serum plasma and cell culture supernatants It uses a quantitative sandwich enzyme immunoassay and color-coded microspheres to measure multiple biomarkers in one well The kit includes microplates detection antibodies standards controls diluents wash solutions substrates and stop solutions for 96 tests With a range of 1 56-400 ng/mL and minimal sample volume it allows rapid reproducible quantification of N-Cadherin across samples