Learn More
Meso Scale Discovery R-PLEX Human NAP-2/CXCL7 ASYQP
SDP
Catalog No. 5000079531
Encompass
The R-PLEX Human NAP-2 (CXCL7) Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of NAP-2 (CXCL7) in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The assay has a detection range of 0 06-1500 pg/mL and requires 25 L sample size per well The total assay time is 3 5 hours This assay allows for the high-throughput sensitive and reproducible quantification of the neutrophil-activating chemokine NAP-2
Catalog No. 50-000-79531 Supplier Meso Scale Discovery Supplier No. K151ACKR22
The R-PLEX Human NAP-2 (CXCL7) Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of NAP-2 (CXCL7) in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The assay has a detection range of 0 06-1500 pg/mL and requires 25 L sample size per well The total assay time is 3 5 hours This assay allows for the high-throughput sensitive and reproducible quantification of the neutrophil-activating chemokine NAP-2