Learn More
Meso Scale Discovery R-PLEX Human NCAM-1 ASY
SDP
Catalog No. 5000082447
Encompass
The R-PLEX Human NCAM-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures NCAM-1 (Neural Cell Adhesion Molecule 1) in human serum plasma and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Calibrator Blend and Sample Diluent The assay has a typical lower limit of detection of 0 05 ng/mL and an upper limit of 500 ng/mL for NCAM-1
Catalog No. 50-000-82447 Supplier Meso Scale Discovery Supplier No. K1517JR2
The R-PLEX Human NCAM-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures NCAM-1 (Neural Cell Adhesion Molecule 1) in human serum plasma and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Calibrator Blend and Sample Diluent The assay has a typical lower limit of detection of 0 05 ng/mL and an upper limit of 500 ng/mL for NCAM-1