Learn More
Meso Scale Discovery R-PLEX Human NCAM-1 ASYQP
SDP
Catalog No. 5000080259
Encompass
The R-PLEX Human NCAM-1 Assay Kit from Meso Scale Discovery measures Neural Cell Adhesion Molecule 1 (NCAM-1) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for simultaneous detection of NCAM-1 and other biomarkers from a small sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay offers a sensitive reproducible method to quantify NCAM-1 levels
Catalog No. 50-000-80259 Supplier Meso Scale Discovery Supplier No. K1517JR22
The R-PLEX Human NCAM-1 Assay Kit from Meso Scale Discovery measures Neural Cell Adhesion Molecule 1 (NCAM-1) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for simultaneous detection of NCAM-1 and other biomarkers from a small sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay offers a sensitive reproducible method to quantify NCAM-1 levels