Learn More
Meso Scale Discovery R-PLEX Human Nephrin ASYQP
SDP
Catalog No. 5000078757
Encompass
The R-PLEX Human Nephrin Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring nephrin levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity a dynamic range of up to 5 logs and low sample volume The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies and necessary calibrators and controls It delivers results in 3 5 hours and contains reagents for one 96-well plate
Catalog No. 50-000-78757 Supplier Meso Scale Discovery Supplier No. K151T4R22
The R-PLEX Human Nephrin Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring nephrin levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity a dynamic range of up to 5 logs and low sample volume The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies and necessary calibrators and controls It delivers results in 3 5 hours and contains reagents for one 96-well plate