Learn More
Meso Scale Discovery R-PLEX Human Neprilysin ASY
SDP
Catalog No. 5000079454
Encompass
The R-Plex Human Neprilysin Assay from Meso Scale Discovery is a 96-well plate assay using electrochemiluminescence (ECL) for quantifying human neprilysin in biological samples It features a pre-coated plate with an ECL label-conjugated antibody specific for neprilysin Samples bind to the immobilized antibody and a ruthenylated detection antibody is added ECL is induced by a read buffer and measured with the MSD instrument with the signal proportional to neprilysin levels The assay offers high sensitivity a wide dynamic range and results in 90 minutes
Catalog No. 50-000-79454 Supplier Meso Scale Discovery Supplier No. K151ABUR2
The R-Plex Human Neprilysin Assay from Meso Scale Discovery is a 96-well plate assay using electrochemiluminescence (ECL) for quantifying human neprilysin in biological samples It features a pre-coated plate with an ECL label-conjugated antibody specific for neprilysin Samples bind to the immobilized antibody and a ruthenylated detection antibody is added ECL is induced by a read buffer and measured with the MSD instrument with the signal proportional to neprilysin levels The assay offers high sensitivity a wide dynamic range and results in 90 minutes