Learn More
Meso Scale Discovery R-PLEX Human Neprilysin ASYQP
SDP
Catalog No. 5000079491
Encompass
The R-PLEX Human Neprilysin Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of human Neprilysin in serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and neprilysin calibrator The kit can test up to 80 samples in duplicate and has a testing time of 3 5 hours Overall this is a high-quality assay kit enabling sensitive and reproducible quantification of human Neprilysin
Catalog No. 50-000-79491 Supplier Meso Scale Discovery Supplier No. K151ABUR22
The R-PLEX Human Neprilysin Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of human Neprilysin in serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and neprilysin calibrator The kit can test up to 80 samples in duplicate and has a testing time of 3 5 hours Overall this is a high-quality assay kit enabling sensitive and reproducible quantification of human Neprilysin