Learn More
Meso Scale Discovery R-PLEX Human Nesfatin-1 ASYQP
SDP
Catalog No. 5000079525
Encompass
The R-PLEX Human Nesfatin-1 Assay is an immunoassay kit for the quantitative determination of Nesfatin-1 in human serum plasma and cell culture supernatants It utilizes a quantitative sandwich enzyme immunoassay technique and takes approximately 3 5 hours to complete The assay has a sensitivity of 0 05 ng/mL and a range of 0 1-1000 ng/mL The kit includes a 96-well strip plate pre-coated with anti-Nesfatin-1 antibody standards detection reagents wash and sample buffers substrate and detailed instructions This assay allows for the specific and sensitive measurement of Nesfatin-1 levels
Catalog No. 50-000-79525 Supplier Meso Scale Discovery Supplier No. K151ABVR22
The R-PLEX Human Nesfatin-1 Assay is an immunoassay kit for the quantitative determination of Nesfatin-1 in human serum plasma and cell culture supernatants It utilizes a quantitative sandwich enzyme immunoassay technique and takes approximately 3 5 hours to complete The assay has a sensitivity of 0 05 ng/mL and a range of 0 1-1000 ng/mL The kit includes a 96-well strip plate pre-coated with anti-Nesfatin-1 antibody standards detection reagents wash and sample buffers substrate and detailed instructions This assay allows for the specific and sensitive measurement of Nesfatin-1 levels