Learn More
Meso Scale Discovery R-PLEX Human NPTX-1 ASYQP
SDP
Catalog No. 5000082224
Encompass
The Meso Scale Discovery R-PLEX Hu Neuronal Pentraxin 1 Assay is a multiplex immunoassay kit for measuring human Neuronal Pentraxin 1 (NP1) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The assay requires 50 L or less per well and processes up to 80 samples per run The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls diluent read buffer and plate seals enabling simple and rapid NP1 quantification
Catalog No. 50-000-82224 Supplier Meso Scale Discovery Supplier No. K151T8R22
Add to Cart
Add to Cart
The Meso Scale Discovery R-PLEX Hu Neuronal Pentraxin 1 Assay is a multiplex immunoassay kit for measuring human Neuronal Pentraxin 1 (NP1) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The assay requires 50 L or less per well and processes up to 80 samples per run The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls diluent read buffer and plate seals enabling simple and rapid NP1 quantification