Learn More
Meso Scale Discovery R-PLEX Human NPY ASY
SDP
Catalog No. 5000079164
Encompass
The R-PLEX Human NPY Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Neuropeptide Y (NPY) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL This product allows for the quantitative measurement of NPY levels from small sample volumes
Catalog No. 50-000-79164 Supplier Meso Scale Discovery Supplier No. K151S0R2
Add to Cart
Add to Cart
The R-PLEX Human NPY Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Neuropeptide Y (NPY) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL This product allows for the quantitative measurement of NPY levels from small sample volumes