Learn More
Meso Scale Discovery R-PLEX Human NT-proBNP ASYQP
SDP
Catalog No. 5000078846
Encompass
The R-PLEX Human IL1RL1/ST2 Assay Kit from Meso Scale Discovery measures IL1RL1/ST2 in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection It offers high sensitivity a 5-log dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies and necessary calibrators and controls The assay takes about 3 hours and is compatible with MSD platforms providing a sensitive and reproducible method for quantifying IL1RL1/ST2 levels
Catalog No. 50-000-78846 Supplier Meso Scale Discovery Supplier No. K1514IR22
The R-PLEX Human IL1RL1/ST2 Assay Kit from Meso Scale Discovery measures IL1RL1/ST2 in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection It offers high sensitivity a 5-log dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies and necessary calibrators and controls The assay takes about 3 hours and is compatible with MSD platforms providing a sensitive and reproducible method for quantifying IL1RL1/ST2 levels