Learn More
Meso Scale Discovery R-PLEX Human Osteocalcin ASY
SDP
Catalog No. 5000080488
Encompass
The Meso Scale Discovery R-PLEX Human Osteocalcin Assay is a multiplex immunoassay panel that measures osteocalcin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay allows for the quantitative measurement of osteocalcin a bone formation biomarker to aid in the evaluation of bone formation turnover and metabolic bone diseases With a short 2 5 hour protocol minimal hands-on time is required
Catalog No. 50-000-80488 Supplier Meso Scale Discovery Supplier No. K151ZIR2
The Meso Scale Discovery R-PLEX Human Osteocalcin Assay is a multiplex immunoassay panel that measures osteocalcin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay allows for the quantitative measurement of osteocalcin a bone formation biomarker to aid in the evaluation of bone formation turnover and metabolic bone diseases With a short 2 5 hour protocol minimal hands-on time is required