Learn More
Meso Scale Discovery R-PLEX Human Osteocalcin ASYQP
SDP
Catalog No. 5000080112
Encompass
The Meso Scale Discovery R-PLEX Human Osteocalcin Assay is a multiplex immunoassay kit for the quantitative measurement of osteocalcin in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the simultaneous measurement of osteocalcin and other biomarkers It includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer sample diluent and wash buffer The total assay time is approximately 3 5 hours
Catalog No. 50-000-80112 Supplier Meso Scale Discovery Supplier No. K151ZIR22
The Meso Scale Discovery R-PLEX Human Osteocalcin Assay is a multiplex immunoassay kit for the quantitative measurement of osteocalcin in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the simultaneous measurement of osteocalcin and other biomarkers It includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer sample diluent and wash buffer The total assay time is approximately 3 5 hours