Learn More
Meso Scale Discovery R-PLEX Human Osteonectin ASY
SDP
Catalog No. 5000078933
Encompass
The R-PLEX Human Osteonectin Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures osteonectin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 8-plex multiplexing and low sample volume requirements This assay allows for the quantitative measurement of osteonectin also known as SPARC or BM-40 which plays a role in bone mineralization and remodeling It requires 50 L of sample and can be read on Meso Scale Diagnostics QuickPlex SQ 120 instrument The R-PLEX assay provides reproducible and reliable quantitation of osteonectin
Catalog No. 50-000-78933 Supplier Meso Scale Discovery Supplier No. K151ZJR2
The R-PLEX Human Osteonectin Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures osteonectin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 8-plex multiplexing and low sample volume requirements This assay allows for the quantitative measurement of osteonectin also known as SPARC or BM-40 which plays a role in bone mineralization and remodeling It requires 50 L of sample and can be read on Meso Scale Diagnostics QuickPlex SQ 120 instrument The R-PLEX assay provides reproducible and reliable quantitation of osteonectin