Learn More
Meso Scale Discovery R-PLEX Human P-Cadherin ASY
SDP
Catalog No. 5000079867
Encompass
The R-Plex Human P-Cadherin Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of P-Cadherin in human serum plasma and cell culture supernatants It utilizes Meso Scale Discovery electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires an MSD instrument and provides components for one 96-well plate including one standard curve comprised of eight points ranging from 3 2 pg/mL to 10 000 pg/mL P-Cadherin
Catalog No. 50-000-79867 Supplier Meso Scale Discovery Supplier No. K151ZLR2
The R-Plex Human P-Cadherin Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of P-Cadherin in human serum plasma and cell culture supernatants It utilizes Meso Scale Discovery electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires an MSD instrument and provides components for one 96-well plate including one standard curve comprised of eight points ranging from 3 2 pg/mL to 10 000 pg/mL P-Cadherin