Learn More
Meso Scale Discovery R-PLEX Human p38 total ASY
SDP
Catalog No. 5000078432
Encompass
The Meso Scale Discovery R-PLEX assay kits are multiplex immunoassays for measuring up to 10 protein biomarkers from one sample Using electrochemiluminescence detection they offer high sensitivity a dynamic range of up to 5 logs and low sample use The kits include plates with capture antibodies detection antibodies calibrators controls buffers and reagents Available for human mouse rat and monkey biomarkers they deliver results in 3 5 hours and are designed for MSD instruments for high performance and ease of use
Catalog No. 50-000-78432 Supplier Meso Scale Discovery Supplier No. K151AKTR2
The Meso Scale Discovery R-PLEX assay kits are multiplex immunoassays for measuring up to 10 protein biomarkers from one sample Using electrochemiluminescence detection they offer high sensitivity a dynamic range of up to 5 logs and low sample use The kits include plates with capture antibodies detection antibodies calibrators controls buffers and reagents Available for human mouse rat and monkey biomarkers they deliver results in 3 5 hours and are designed for MSD instruments for high performance and ease of use