Learn More
Meso Scale Discovery R-PLEX Human PC9/PCSK9 ASY
SDP
Catalog No. 5000078922
Encompass
The R-PLEX Human PC9/PCSK9 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously measures Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9) and Progranulin (PC9) in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of PCSK9 and PC9 levels from a single small-volume sample With a broad dynamic range and rapid protocol this assay kit allows high-throughput profiling of PCSK9 and PC9 biomarkers involved in lipid metabolism and inflammation
Catalog No. 50-000-78922 Supplier Meso Scale Discovery Supplier No. K151ABAR2
The R-PLEX Human PC9/PCSK9 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously measures Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9) and Progranulin (PC9) in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of PCSK9 and PC9 levels from a single small-volume sample With a broad dynamic range and rapid protocol this assay kit allows high-throughput profiling of PCSK9 and PC9 biomarkers involved in lipid metabolism and inflammation