Learn More
Meso Scale Discovery R-PLEX Human PDGF-A ASY
SDP
Catalog No. 5000078533
Encompass
The Meso Scale Discovery R-PLEX Human PDGF-AA Assay Kit is a multiplex immunoassay that measures PDGF-AA in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity and broad dynamic range The assay requires 50 L or less sample volume and takes 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The kit has high specificity and low cross-reactivity It allows simultaneous measurement of multiple analytes from a single sample
Catalog No. 50-000-78533 Supplier Meso Scale Discovery Supplier No. K151H1R2
The Meso Scale Discovery R-PLEX Human PDGF-AA Assay Kit is a multiplex immunoassay that measures PDGF-AA in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity and broad dynamic range The assay requires 50 L or less sample volume and takes 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The kit has high specificity and low cross-reactivity It allows simultaneous measurement of multiple analytes from a single sample