Learn More
Meso Scale Discovery R-PLEX Human PDGF-A ASYQP
SDP
Catalog No. 5000082096
Encompass
The R-PLEX Human PDGF-AA Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures PDGF-AA in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can assay up to 80 samples in duplicates in 3 5 hours The lower limit of detection is 1 5 pg/mL
Catalog No. 50-000-82096 Supplier Meso Scale Discovery Supplier No. K151H1R22
The R-PLEX Human PDGF-AA Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures PDGF-AA in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can assay up to 80 samples in duplicates in 3 5 hours The lower limit of detection is 1 5 pg/mL