Learn More
Meso Scale Discovery R-PLEX Human PDGF-B ASY
SDP
Catalog No. 5000079596
Encompass
The R-PLEX Human PDGF-B Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures PDGF-B levels in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity and a broad dynamic range This quantitative assay has high reproducibility and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibration standard and sample diluent It can accurately quantify PDGF-B levels between 1 5-1000 pg/mL The assay takes 3 5 hours to complete
Catalog No. 50-000-79596 Supplier Meso Scale Discovery Supplier No. K151X0R2
The R-PLEX Human PDGF-B Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures PDGF-B levels in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity and a broad dynamic range This quantitative assay has high reproducibility and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibration standard and sample diluent It can accurately quantify PDGF-B levels between 1 5-1000 pg/mL The assay takes 3 5 hours to complete