Learn More
Meso Scale Discovery R-PLEX Human PDGF-C ASY
SDP
Catalog No. 5000078853
Encompass
The R-PLEX Human PDGF-C Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures PDGF-C levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-78853 Supplier Meso Scale Discovery Supplier No. K151ABCR2
The R-PLEX Human PDGF-C Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures PDGF-C levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL