Learn More
Meso Scale Discovery R-PLEX Human PDGF-C ASYQP
SDP
Catalog No. 5000079449
Encompass
The R-PLEX Human PDGF-C Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures PDGF-C levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator diluent and lyophilized PDGF-C calibrator The assay can accurately quantify PDGF-C concentrations between 1 53 pg/mL to 2500 pg/mL
Catalog No. 50-000-79449 Supplier Meso Scale Discovery Supplier No. K151ABCR22
The R-PLEX Human PDGF-C Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures PDGF-C levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator diluent and lyophilized PDGF-C calibrator The assay can accurately quantify PDGF-C concentrations between 1 53 pg/mL to 2500 pg/mL