Learn More
Meso Scale Discovery R-PLEX Human PR/PGR ASY
SDP
Catalog No. 5000080498
Encompass
The R-PLEX Human PR/PGR Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures Progesterone Receptor (PR) and Progesterone (PGR) in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SECTOR Imager and provides sensitive accurate and reproducible quantitation of PR and PGR levels With a broad dynamic range and rapid protocol this assay allows high-throughput profiling of PR and PGR useful for drug discovery and biomarker research The R-PLEX panel provides an efficient alternative to ELISA with better sensitivity wider dynamic range and multiplexing capabilities up to 10-plex
Catalog No. 50-000-80498 Supplier Meso Scale Discovery Supplier No. K151AHXR2
The R-PLEX Human PR/PGR Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures Progesterone Receptor (PR) and Progesterone (PGR) in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SECTOR Imager and provides sensitive accurate and reproducible quantitation of PR and PGR levels With a broad dynamic range and rapid protocol this assay allows high-throughput profiling of PR and PGR useful for drug discovery and biomarker research The R-PLEX panel provides an efficient alternative to ELISA with better sensitivity wider dynamic range and multiplexing capabilities up to 10-plex