Learn More
Meso Scale Discovery R-PLEX Human PR/PGR ASYQP
SDP
Catalog No. 5000079131
Encompass
The R-Plex Human PR/PGR Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures the levels of progesterone receptor (PR) and progesterone (PGR) in human serum plasma and cell culture supernatants This 10-plex assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can be run on Meso Scale Diagnostic s QuickPlex SQ 120 instrument and MESO QuickPlex SQ 120 Plates in as little as 3 hours This assay allows for the quantitative measurement of PR and PGR levels which can aid in monitoring hormone therapy and diagnosing hormone-related disorders
Catalog No. 50-000-79131 Supplier Meso Scale Discovery Supplier No. K151AHXR22
The R-Plex Human PR/PGR Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures the levels of progesterone receptor (PR) and progesterone (PGR) in human serum plasma and cell culture supernatants This 10-plex assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can be run on Meso Scale Diagnostic s QuickPlex SQ 120 instrument and MESO QuickPlex SQ 120 Plates in as little as 3 hours This assay allows for the quantitative measurement of PR and PGR levels which can aid in monitoring hormone therapy and diagnosing hormone-related disorders