Learn More
Meso Scale Discovery R-PLEX Human PRDX6 ASY
SDP
Catalog No. 5000080284
Encompass
The R-PLEX Human PRDX6 Assay Kit from Meso Scale Discovery is used to quantitatively measure peroxiredoxin-6 (PRDX6) in human serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of PRDX6 The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent Results are obtained in as little as 3 5 hours using an MSD instrument capable of reading electrochemiluminescent signals like the MESO QuickPlex SQ 120
Catalog No. 50-000-80284 Supplier Meso Scale Discovery Supplier No. K1511NR2
The R-PLEX Human PRDX6 Assay Kit from Meso Scale Discovery is used to quantitatively measure peroxiredoxin-6 (PRDX6) in human serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of PRDX6 The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent Results are obtained in as little as 3 5 hours using an MSD instrument capable of reading electrochemiluminescent signals like the MESO QuickPlex SQ 120