Learn More
Meso Scale Discovery R-PLEX Human PRDX6 ASYQP
SDP
Catalog No. 5000078849
Encompass
The R-PLEX Human PRDX6 Assay Kit from Meso Scale Discovery is a 96-well plate assay that measures PRDX6 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This quantitative immunoassay contains a plate coated with capture antibody detection antibody and standards to generate a standard curve The kit includes sufficient reagents for one 96-well plate and can test up to 40 samples in duplicate Assay time is less than 3 5 hours
Catalog No. 50-000-78849 Supplier Meso Scale Discovery Supplier No. K1511NR22
Add to Cart
Add to Cart
The R-PLEX Human PRDX6 Assay Kit from Meso Scale Discovery is a 96-well plate assay that measures PRDX6 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This quantitative immunoassay contains a plate coated with capture antibody detection antibody and standards to generate a standard curve The kit includes sufficient reagents for one 96-well plate and can test up to 40 samples in duplicate Assay time is less than 3 5 hours