Learn More
Meso Scale Discovery R-PLEX Human Procalcitonin ASY
SDP
Catalog No. 5000081620
Encompass
The R-PLEX Human Procalcitonin Assay from Meso Scale Diagnostics is a quantitative electrochemiluminescent immunoassay for the in vitro diagnostic measurement of procalcitonin in human serum plasma and whole blood This assay utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The assay has a measuring range of 0 02 - 75 ng/mL PCT with an LLOQ of 0 02 ng/mL The total assay time is approximately 3 5 hours This assay provides a precise accurate method to aid in the risk assessment of critically ill patients for progression to severe sepsis and septic shock
Catalog No. 50-000-81620 Supplier Meso Scale Discovery Supplier No. K151AJTR2
The R-PLEX Human Procalcitonin Assay from Meso Scale Diagnostics is a quantitative electrochemiluminescent immunoassay for the in vitro diagnostic measurement of procalcitonin in human serum plasma and whole blood This assay utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The assay has a measuring range of 0 02 - 75 ng/mL PCT with an LLOQ of 0 02 ng/mL The total assay time is approximately 3 5 hours This assay provides a precise accurate method to aid in the risk assessment of critically ill patients for progression to severe sepsis and septic shock