Learn More
Meso Scale Discovery R-PLEX Human Progranulin ASY
SDP
Catalog No. 5000080494
Encompass
The Meso Scale Discovery R-PLEX Human Progranulin Assay is a multiplex immunoassay panel that measures Progranulin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 8-plex multiplexing and low sample volume requirements This assay allows for the quantitative measurement of Progranulin which is involved in wound healing inflammation and neuronal survival It requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Instruments The panel contains a plate coated with capture antibodies detection antibodies calibrators and controls
Catalog No. 50-000-80494 Supplier Meso Scale Discovery Supplier No. K151ALER2
The Meso Scale Discovery R-PLEX Human Progranulin Assay is a multiplex immunoassay panel that measures Progranulin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 8-plex multiplexing and low sample volume requirements This assay allows for the quantitative measurement of Progranulin which is involved in wound healing inflammation and neuronal survival It requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Instruments The panel contains a plate coated with capture antibodies detection antibodies calibrators and controls