Learn More
Meso Scale Discovery R-PLEX Human Progranulin ASYQP
SDP
Catalog No. 5000078431
Encompass
The Meso Scale Discovery R-PLEX Human Progranulin Assay is a multiplex immunoassay panel that measures Progranulin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 8-plex multiplexing and low sample volume requirements This assay allows for the quantitative measurement of Progranulin which is involved in wound healing inflammation and neuronal survival It requires 50 L of sample and can perform up to 80 tests per kit The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78431 Supplier Meso Scale Discovery Supplier No. K151ALER22
The Meso Scale Discovery R-PLEX Human Progranulin Assay is a multiplex immunoassay panel that measures Progranulin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 8-plex multiplexing and low sample volume requirements This assay allows for the quantitative measurement of Progranulin which is involved in wound healing inflammation and neuronal survival It requires 50 L of sample and can perform up to 80 tests per kit The assay takes approximately 3 5 hours to complete