Learn More
Meso Scale Discovery R-PLEX Human proMMP-10 ASY
SDP
Catalog No. 5000078506
Encompass
The R-Plex Human ProMMP-10 Assay is a multiplex immunoassay kit from Meso Scale Discovery that enables the simultaneous quantitative measurement of ProMMP-10 protein in human serum plasma urine cell culture supernatant and other biological fluids This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator This assay can be used for biomarker discovery and validation With minimal hands-on time required it provides rapid and reliable quantification of ProMMP-10
Catalog No. 50-000-78506 Supplier Meso Scale Discovery Supplier No. K151ALPR2
The R-Plex Human ProMMP-10 Assay is a multiplex immunoassay kit from Meso Scale Discovery that enables the simultaneous quantitative measurement of ProMMP-10 protein in human serum plasma urine cell culture supernatant and other biological fluids This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator This assay can be used for biomarker discovery and validation With minimal hands-on time required it provides rapid and reliable quantification of ProMMP-10