Learn More
Meso Scale Discovery R-PLEX Human proMMP-10 ASYQP
SDP
Catalog No. 5000082015
Encompass
The R-Plex Human ProMMP-10 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of proMMP-10 protein in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be fully automated and has a rapid 2 5 hour turnaround time It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and lyophilized calibrator
Catalog No. 50-000-82015 Supplier Meso Scale Discovery Supplier No. K151ALPR22
The R-Plex Human ProMMP-10 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of proMMP-10 protein in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be fully automated and has a rapid 2 5 hour turnaround time It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and lyophilized calibrator